ContactUsLinkComponent
PCR plate, 384-well, PP, 0,03 ml, full skirted, low profile, BIO-CERT® PCR QUALITY
PCR plate, 384-well, PP, 0,03 ml, full skirted, low profile, BIO-CERT® PCR QUALITY
Catalog # 732-3977
Supplier:  Brand
undefined
PCR plate, 384-well, PP, 0,03 ml, full skirted, low profile, BIO-CERT® PCR QUALITY
Catalog # 732-3977
Supplier:  Brand
Specifications
  • Frame colour:
    Transparent
  • No. of wells:
    384
  • Profile:
    Low
  • Skirt:
    Fully skirted
  • Well colour:
    Transparent
  • Pk:
    50
  • Well volume:
    0,03 ml
Specifications
About this item
These 384-well PCR plates are an economical solution for automatic high-throughput analysis.
  • For applications in automation and high-throughput analysis
  • Rapid heat transfer through thin vessel walls
  • Can be sealed reliably with self-adhesive sealing films
The thin walls allow fast transmission of the temperature specified by the cycler, thus reducing cycle times. The stable design of the plates makes them ideal for automation, as they can be gripped by robot systems without twisting.
To minimise evaporation with small sample volumes up to 2 µl and to prevent contamination, seal the plates with the appropriate sealing film. The cleanroom quality of the PCR consumables, including 384-well PCR plates from BRAND guarantees reliable results. They are free of DNase, DNA, RNase, and endotoxins.