Order Entry
Austria
ContactUsLinkComponent
VWR®, Nominal flash point standard (65 °C)
VWR®, Nominal flash point standard (65 °C)
Catalog # 91688.230
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Nominal flash point standard (65 °C)
Catalog # 91688.230
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • CAS No.:
    1120-21-4
  • Description:
    Nominal flash point standard (65 °C)
  • Formula:
    C₁₁H₂₄
  • Storage temperature:
    Ambient (>5 °C)
  • Description:
    Nominal Flash Point Standard (65°C)

Specifications

About this item

Caution: Carcinogen.