
Ethanol 20% for biopharmaceutical production
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Density:0.9686 g/cm³ (20°C)
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,III
REACH:01-2119457610-43-XXXX
:GHS07
Specification Test Results
- assay:19,5 - 20,5%
- appearance:passes test
- evaporation residue:≤ 10 pmm
- residual solvents (ICHQ3C):excluded by manufacturing process
- residual metal catalysts:unlikely to be present
- endotoxin:≤ 2,5 EU/ml
- TAMC:≤ 10 CFU/ml
- filtered through 0,2µm:confirmed
Specification Test Results
Related Information
Certificates
Search by Lot NumberEthanol 20% for Biopharmaceutical Production
Additional Product Details
spec_VWRC85901.290 spec_VWRC85901.410 spec_VWRC85901.550
Product specification
Frequently Bought Together