Safety helmet chinstraps
Artikelnr. MMMAX5-P4PTCS1
Lieferant: 3M
Spezifikationen
- Description:Safety helmet chinstraps
- Pk:1
Spezifikationen
Über diesen Artikel
This four-point chinstrap features a magnetic buckle and fits the 3M™ SecureFit™ Safety Helmet series X5000 and X5500.
- Magnetic buckle allows for single-handed fastening and opening
- Easy and quick to use
This four-point chinstrap is designed with quick and smooth adjustment features for a custom fit.