Orders LinkContactUsLinkComponent
Lightweight Plastic Pallets
 :   MULTIROIR
Lightweight Plastic Pallets
 :  MULTIROIR
Lightweight Plastic Pallets
Lightweight Plastic Pallets
Lightweight plastic pallets are made from recycled PP or HDPE and are a real alternative to wooden pallets.
  • Material: Recycled PP
  • Security lip 6 mm
  • Temperature from −20 to +40 °C
  • Open deck
These pallets are both lightweight and sturdy. They are available in sizes 120×80 and 120×100 with 9 feet or 3 skids. The 9 feet pallets are nestable and stackable.
Product Family Options
  • Description
    W×D×H ext.
    Colour
  • 9 feet on base, 850 kg max. dynamic load capacity, 1200 kg max. static load capacity
    1200×800×134 mm
    Black
    1 * 1 ST
    1 * 1 ST - 50,20 €
    50,20 €
    1 * 1 ST
       50,20 €
  • 3 skids, 1000 kg max. dynamic load capacity, 2000 kg max. static load capacity
    1200×800×150 mm
    Black
    1 * 1 ST
    1 * 1 ST - 87,60 €
    87,60 €
    1 * 1 ST
       87,60 €
  • 9 feet on base, 850 kg max. dynamic load capacity, 1500 kg max. static load capacity
    1200×1000×134 mm
    Black
    1 * 1 ST
    1 * 1 ST - 70,20 €
    70,20 €
    1 * 1 ST
       70,20 €
  • 3 skids, 1000 kg max. dynamic load capacity, 2000 kg max. static load capacity
    1200×1000×150 mm
    Black
    1 * 1 ST
    1 * 1 ST - 97,60 €
    97,60 €
    1 * 1 ST
       97,60 €