ContactUsLinkComponent
Powered air respirator headtop collar
Powered air respirator headtop collar
Catalog # MMMAM444
Supplier:  3M
undefined
Powered air respirator headtop collar
Catalog # MMMAM444
Supplier:  3M
Specifications
  • Description:
    Powered air respirator headtop collars
  • Model:
    M-444
  • Pk:
    1
Specifications
About this item
Replacement inner collar for use with the Versaflo™ M-400 Headtop. The inner collar zips into the shroud and provides a comfortable respiratory seal.
  • Replacement inner collar
  • Provides a comfortable respiratory seal
  • Made from polypropylene coated knitted polyamid
Replacement inner collar is designed for use with the 3M™Versaflo™ M-400 Series Helmets.