ContactUsLinkComponent
Cleaning solution for ultrapure water systems, Arium®
Cleaning solution for ultrapure water systems, Arium®
Catalog # 171-1024
Supplier:  Sartorius
Cleaning solution for ultrapure water systems, Arium®
Catalog # 171-1024
Supplier:  Sartorius
Specifications
  • Description:
    Arium® cleaning solution, 50 ml syringe, sodium hypochlorite, white
  • Seq:
    22
  • Pk:
    1
Specifications
About this item
The Arium® cleaning solution is used to effectively remove impurities that can built up over time during water treatment.
  • Highly effective cleaning
  • Free of organic components
  • Gentle to the materials
Regular cleaning not only removes impurities but also leads to a longer service life of consumables and increases the flow rate.
Filled and ready to use in the practical cleaning syringe, the cleaning solution can be used directly.