Disposable flat bottom culture tubes, screw cap style, bulk pack, PYREX®
Supplier: Corning
Certificates
About this item
Tube has flat bottom and comes without marking spot. Tubes are made of borosilicate glass and are sold without caps.
Specifications
- Catalog No:
- 734-4237
- 734-4238
- 734-4236
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey