ContactUsLinkComponent
96-Well high content screening microplates with film bottom, Corning®
96-Well high content screening microplates with film bottom, Corning®
Catalog # 734-2763
Supplier:  Corning
   Low Manufacturing Impact
undefined
96-Well high content screening microplates with film bottom, Corning®
Catalog # 734-2763
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    96
  • Sterile:
    +
  • Colour:
    Black/clear
  • Description:
    Half area microplate, with lid, tissue culture treated
  • Seq:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Pk:
    16
  • Well volume:
    200 µl
Specifications
About this item
With an ultraclear film bottom and excellent flatness (whole plate and intra-well), these microplates are ideal for high resolution cellular imaging applications.
  • TC-treated, flat well bottom black microplate
  • Made of cyclic olefin copolymer
  • Ultraclear 190 µm film bottom thickness for imaging microscopy applications
  • Low background fluorescence and minimal crosstalk allows highest possible optical quality for cell based assays
  • Half area 96-well microplate reduces reagent consumption
  • Sterile
  • Inter and inter-well film bottom flatness optimised for high content applications
The microplate and film are manufactured from cyclic olefin copolymer, which has excellent optical properties, chemical resistance, and mechanical stability.