Overalls with hood, AlphaTec® MICROGARD 2300
: Ansell
Medium-duty chemical barrier, Type 5/6 protection.
- A good protective barrier to numerous inorganic liquid chemicals including acids and bases
- Lightweight, yet relatively strong and durable
- Standard protective features include respirator fit hood and a zip flap with self-adhesive tape closure
- Highly visible bright yellow for improved worker safety
- Anti-static tested according to EN 1149-5
MICROGARD 2300 fabric is comprised of a polyethylene (PE) barrier coating with a bi-component non-woven inner layer, the combination of which provides an excellent barrier to many harmful chemicals, while remaining lightweight and relatively strong and durable. Stitched and taped seems provide superior strength, as well as a liquid and particle barrier.
Coveralls feature a 3-piece hood, elasticated wrists, waist, and ankles, a 2-way front zipper with resealable storm flap, and finger loops.
- Catalog No:
- ANSEYY23B-00111-07
- ANSEYY23B-00111-06
- ANSEYY23B-00111-05
- ANSEYY23B-00111-04
- ANSEYY23B-00111-03
- ANSEYY23B-00111-02
- ANSEYY23B-00111-09
- ANSEYY23B-00111-08
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
Frequently Bought Together