Chemical resistant gloves, AlphaTec® 58-001 ESD
: Ansell
AlphaTec® 58-001 ESD gloves provide advanced chemical protection in mitigating static buildup across surface areas, specifically when working with chemicals and liquids in environments with static hazards or high explosive risks.
- Unique nitrile and carbon formulation consistently dissipates static across the surface and through the glove
- Nitrile compound offers far superior snag, puncture, and abrasion protection compared to rubber or neoprene
- Reverse lozenge palm finish for improved dry/wet grip
- Foldable cuff gutter prevents dripping on the forearm
- Anti-static flocking layer for ease of donning/doffing and moisture management
Nitrile barrier provides advanced chemical protection from many frequently used chemicals. The AlphaTec 58-001 series offers Type A chemical protection under new EN ISO 374:2016. Unique Nitrile composition allows effective dissipation of static build-up.
: EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
Food contact approved
Food contact approved
- Catalog No:
- 112-6419
- 112-6415
- 112-6416
- 112-6418
- 112-6417
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Lozenge
- Lozenge
- Lozenge
- Lozenge
- Lozenge
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Palm thickness:
- 0,38 mm
- 0,38 mm
- 0,38 mm
- 0,38 mm
- 0,38 mm
- EN norm:
- EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
- EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
- EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
- EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
- EN ISO 374-1:2016 Type A (AJKLOPT), EN ISO 374-5 VIRUS, EN 388:2016 (4101X), EN 1149, EN 16350
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- Food contact approved:
- +
- +
- +
- +
- +
Frequently Bought Together