HyFlex® 11-754 Cut-Resistant Industrial Gloves with INTERCEPT™ Technology
Supplier: Ansell
Certificates
About this item
Ansell's lightest ANSI A4/ISO Level D cut resistant touchscreen glove.
- Touchscreen compatible to ensure that workers do not need to change or remove gloves between tasks
- Designed to fit the natural shape of the hand to help reduce hand fatigue
- Clean and skin friendly; dermatologically approved
Lightest weight ANSI A4/ISO Cut D glove featuring INTERCEPT™ Technology, these ultra-thin breathable gloves offer high cut resistance, while maintaining enhanced flexibility and tactility. Touchscreen compatible glove for enhanced productivity. Ultra-lightweight design provides dexterity for handling and assembly of sharp parts.
Specifications
- Catalog No:
- ANSE11-754/10
- ANSE11-754/12
- ANSE11-754/11
- ANSE11-754/9
- ANSE11-754/6
- ANSE11-754/8
- ANSE11-754/7
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Gauge:
- 18
- 18
- 18
- 18
- 18
- 18
- 18
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- +
- +