About this item
Fully coated in nitrile, EDGE® 48-500 cut and sewn heavy-duty gloves offer a good level of resistance to abrasive materials and hazards for handling heavy-duty tasks.
- Good level of abrasion protection
- Full nitrile coating delivers long-lasting performance
- Good barrier to oils, greases and liquids
- FDA-approved materials: Safe for food contact and packaging
A cost-effective solution for heavy weight applications requiring a good level of abrasion resistance, along with liquid repellency. Nitrile dipped cut and sew glove for heavy handling, offering good resistance to abrasive material. Stronger and more flexible than PVC, while providing a good barrier to grease and oil. Will not degrade like leather or latex. The economical choice for a heavy-duty glove with quality features.
Recommended for: Cold plastic moldings; heavy-duty cables; rough casting and cores; steel bars, sheet metals, cans and other metal products; street cleaning and refuse collection.
Specifications
- Catalog No:
- ANSE48-500/9
- ANSE48-500/8
- ANSE48-500/10
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- Fully nitrile coated
- Fully nitrile coated
- Fully nitrile coated
- Cuff style:
- Safety cuff
- Safety cuff
- Safety cuff
- EN norm:
- EN 388:2016 (4111B)
- EN 388:2016 (4111B)
- EN 388:2016 (4111B)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +