Orders LinkContactUsLinkComponent
Spinner flasks with solid cap, disposable, Corning®
Spinner flasks with solid cap, disposable, Corning®
Supplier:   Corning
Spinner flasks with solid cap, disposable, Corning®
Supplier:  Corning
Certificates
Spinner flasks with solid cap, disposable, Corning®
Spinner flasks with solid cap, disposable, Corning®
4 Options Available Below
Product Details & Documents
The Corning 1 L spinner flask body is constructed of polystyrene, paddle and shaft from polypropylene and caps from high density polyethylene. All are made of USP Class VI and ISO 10993-18 compliant materials
  • Recommended working range of 300 ml to 1 L
  • Extended paddle design ensures constant top-to-bottom motion of the liquid without having to adjust the height of the paddle
  • Bottom contours on the paddle hug the shape of the flask, eliminating dead spots
  • Two wide neck openings for easy pouring; easily accept a 100 ml pipet
  • Vertical side baffles break up laminar flow - ensures better mixing and maximises cell production
  • Each system is supplied individually double bagged
  • Sterilised by gamma irradiation; have the highest sterility assurance level (SAL) of 10⁻⁶ and meets USP XXIV:71 bacteriostasis, fungistasis and sterility standards
Ordering information: Single use only. Flasks cannot be autoclaved or resterilised with alcohol. Any attempt to do so will result in degradation of the materials. Optional vented caps (Cat. No 3567) and Aseptic transfer caps (Cat. No. 3562) are available Designed for both suspension and adherent cell culture applications and optimised for use with corning microcarriers
Product Family Options
Product Information
  • Capacity
    Neck
    No. of side arms
    Recommended working volume
    Ø×H
    Availability
    Price
  • 734-4103
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 2 700,00 €
    2 700,00 €
    1 * 6 ST
       2 700,00 €
  • 392-0308
    Flasks with one solid cap and one aseptic transfer cap on side arms*
    Flasks with one solid cap and one aseptic transfer cap on side arms*
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 2 710,00 €
    2 710,00 €
    1 * 4 ST
       2 710,00 €
  • 392-0398
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    1000 ml
    side arm necks thread GL 45
    2
    300-1000 ml
    137×245 mm
    Log in for availability
    Log in for availability
    1 * 6 ST
    1 * 6 ST - 2 700,00 €
    2 700,00 €
    1 * 6 ST
       2 700,00 €
  • 392-0399
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    Flasks, pre-assembled, with one solid cap and one aseptic transfer cap on side arms*
    3000 ml
    side arm necks thread GL 45
    2
    1000-3000 ml
    188×259 mm
    Log in for availability
    Log in for availability
    1 * 4 ST
    1 * 4 ST - 2 710,00 €
    2 710,00 €
    1 * 4 ST
       2 710,00 €
Accessories
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    Vent cap, 0,2 μm vent, for 500, 1000 and 3000 ml spinner flasks
    734-4102
    GL45
    Log in for availability
    1 * 4 ST - 44,90 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 3000 ml spinner flask
    734-4100
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    Flat cap, PE, disposable, orange, for 500, 1000 and 3000 ml spinner flasks
    CORN3568
    Log in for availability
    1 * 4 ST - 41,20 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 500 ml spinner flask
    734-4101
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €
Product Information
  • Catalog #
    Thread
    Availability
    Price
  • Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    Side arm aseptic transfer cap, dip tube with 0,2 μm vent, male luer lock, for 1000 ml spinner flask
    734-4098
    GL45
    Log in for availability
    1 * 2 ST - 449,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    Modified impeller assembly for magnetically-driven 36 L Corning® Proculture® spinner flask, reusable
    CORN401901
    Log in for availability
    1 * 1 ST - 1 340,00 €