ContactUsLinkComponent
96-well polyvinyl chloride (PVC) microplates
The site is currently experiencing technical difficulties. Please retry later. We apologize for any inconvenience.
96-well polyvinyl chloride (PVC) microplates
Artikelnr. 734-1517
Supplier:  Corning
undefined
96-well polyvinyl chloride (PVC) microplates
Artikelnr. 734-1517
Supplier:  Corning
Specifications
  • Recommended working volume:
    50 - 150 µl
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Round bottom PVC microplates, untreated
  • Pk:
    100
Specifications
About this item
Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 µl (260 µl for V-bottom)
  • Working well volume of 50 µl to 150 µl
Ordering information: Lids are not available.