Order Entry
Austria
ContactUsLinkComponent
Portable fluorometer with Bluetooth, DR1300 FL
Portable fluorometer with Bluetooth, DR1300 FL
Catalog # 705-1194
Supplier:  Hach
undefined
Portable fluorometer with Bluetooth, DR1300 FL
Catalog # 705-1194
Supplier:  Hach

Specifications

  • Detector:
    Silicon photodiode
  • Light source:
    UV LED, 365 nm
  • Memory:
    16 GB SD card or 60000000 data points
  • Description:
    DR1300 FL portable fluorometer (Bluetooth)
  • W×D×H:
    265×62×88 mm
  • Output:
    Bluetooth
  • Weight:
    0,6 kg
  • Pk:
    1

Specifications

About this item

Portable lab metre for ultra low range chlorine and sulphite detection.

  • Provides precise and accurate results
  • Tests are easy-to-perform, with ULR analysis in minutes
  • Reduce biofouling potential
  • Save on dechlorination costs
  • Handles traditional interferences with ease
  • Bluetooth capability allows easy data transfer
  • Decrease in toxic reagents and chemical waste (as detector chemical addition is a fraction of traditional tests)
  • Indoor or outdoor use
  • Graphical LCD with backlight, 160×240 pixels
  • Hach fluorescence measurement method
  • 12 months warranty
  • Parameters include Chlorine free and total, Sulphite

The DR1300 FL portable fluorometer instrument, combined with specially formulated reagents (not included) create a fluorescence that indicates chlorine at ultra low ranges. DR1300 FL offers the most simple way to be confident that chlorine has been removed or reduced from your process.

With this knowledge, you’ll gain the insights you need to manage your dechlorination process and avoid chlorine damage to reverse osmosis (RO) membranes and other valuable assets.

Measure free and total chlorine plus sulphite in the low ppb range:
Free Chlorine: 2 to 100 µg/L (ppb) Cl₂; Total Chlorine: 3 to 100 µg/L (ppb) Cl₂; and Sulphite: 6 to 500 µg/L (ppb) SO₃.

Operating temperature: 4 to 49 °C (40 to 120 °F), 0 to 85% relative humidity (non-condensing)
Power supply: 4 AA alkaline batteries
Wavelength accuracy: ±1 nm

Certifications: DoC CE. IP 65 enclosure rating.
Ordering information: Supplied with DR1300 FL fluorometer, 4× AA batteries, sample vial adapter, sample vial adapter cover, USB Bluetooth converter and user manual. Reagents are not included and have to be purchased separately. Contact your Avantor representative for more information.

Cap

Product Information