ContactUsLinkComponent
Water stills, double distillation, Puridest
 :  LAUDA
undefined
Water stills, double distillation, Puridest
 :  LAUDA
  • Catalog No:
  • 171-1315
  • 171-1322
  • 171-1323
  • 171-1321
  • 171-1332
  • 171-1312
  • 171-1331
  • 171-1334
  • 171-1313
  • Output:
  • 4 L/h
  • 8 L/h
  • 4 L/h
  • 4 L/h
  • 8 L/h
  • 4 L/h
  • 4 L/h
  • 4 L/h
  • 8 L/h
  • Conductivity:
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • 2,2 µS/cm (conductivity stage 1/25 °C); 1,6 µS/cm (conductivity stage 2/25 °C)
  • Ambient temperature:
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • 10...40 °C
  • Power:
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • 400 V; 3/N/PE; 50/60 Hz
  • W×D×H:
  • 650×365×390 mm
  • 700×390×700 mm
  • 650×365×390 mm
  • 550×280×570 mm
  • 700×390×700 mm
  • 550×280×570 mm
  • 550×280×570 mm
  • 650×365×390 mm
  • 700×390×700 mm
  • Weight:
  • 28 kg
  • 45 kg
  • 28 kg
  • 28 kg
  • 45 kg
  • 28 kg
  • 28 kg
  • 26 kg
  • 48 kg
Frequently Bought Together