Order Entry
Austria
ContactUsLinkComponent
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Supplier:  VWR Seradigm
Certificates
undefined
Avantor® Seradigm, Select Grade Fetal Bovine Serum (FBS)
Supplier:  VWR Seradigm
Certificates

Specifications

  • Catalog No:
  • SERD76419-586
  • 1300-500CGH
  • 1300-500CH
  • 1300-500CG
  • SERD76419-590
  • 1300-500CR
  • Endotoxin:
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • ≤10 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Frozen
  • Frozen
  • Frozen
  • Frozen
  • Frozen
  • Frozen

Specifications

Related Information

Frequently Bought Together