Order Entry
Austria
ContactUsLinkComponent
SPINeasy DNA/RNA/protein all in one kit, 50 preparations, MP Biomedicals
SPINeasy DNA/RNA/protein all in one kit, 50 preparations, MP Biomedicals
Catalog # 116544050.
Supplier:  MP Biomedicals
CAS Number:  
SPINeasy DNA/RNA/protein all in one kit, 50 preparations, MP Biomedicals
Catalog # 116544050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    50 Tests
  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total RNA
  • Sample type:
    Human/animal cells and tissues
  • Target molecule:
    Co-purification of RNA, DNA and protein
  • Description:
    SPINeasy DNA/RNA/protein all-in-one kit
  • Seq:
    02

Specifications

About this item

Quick and efficient isolation of DNA, RNA and protein from the same sample using spin column technology.

  • Rapidly and efficiently isolates DNA, RNA and proteins from a single sample
  • Lysing Matrix tubes are supplied for thorough sample lysis by bead beating
  • High binding silica spin columns allow an easy DNA and RNA extraction process
  • Top quality DNA, RNA and proteins are ready for downstream sensitive applications
  • No hazardous chemicals

SPINeasy DNA/RNA/Protein All-In-One Kit utilizes a convenient workflow and silica-membrane spin-columns to isolate without the use of toxic substances such as phenol and chloroform DNA, RNA and protein components from the same sample.

The use of our specially formulated Lysis Buffer R and Lysing Matrix A in combination with FastPrep® Instruments from MP Biomedicals enables highly efficient lysis of tissue samples within seconds. DNA, RNA and proteins are then sequentially purified from the same lysate. Briefly, DNA is adsorbed onto the first spin column, while the flow-through is collected and used for RNA purification.

The second spin column captures RNA and the flow-through from this step is used for protein extraction. DNA and RNA are bound on the first and second columns, respectively, and then washed and eluted.

In the final extraction, proteins are precipitated out of solution, pelleted down by centrifugation and washed and resuspended. Each molecular component is then immediately available for their respective downstream applications.