Order Entry
Austria
ContactUsLinkComponent
DL-α-Tocopherol acetate USP
DL-α-Tocopherol acetate USP
Catalog # SPCMVI140-100ML
Supplier:  Spectrum Chemical
CAS Number:  
DL-α-Tocopherol acetate USP
Catalog # SPCMVI140-100ML
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    96% Min.

Specification Test Results

About this item

 :
 :472.75 g/mol
Density:0.96
Storage Temperature:Ambient
EINECS:231-710-0
CAS Number:7695-91-2
 :52225-20-4
Formula:C₃₁H₅₂O₃
MDL Number:MFCD00072042