Order Entry
Austria
ContactUsLinkComponent
Calcium chloride, anhydrous ≥93.0%, pellets FCC
Calcium chloride, anhydrous ≥93.0%, pellets FCC
Catalog # SPCMC1977-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Calcium chloride, anhydrous ≥93.0%, pellets FCC
Catalog # SPCMC1977-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    93.0 - 100.5 %
  • Acid-Insoluble Substances:
    max. 0.02 %
  • Arsenic (As):
    max. 3 mg/kg
  • Fluoride:
    max. 0.004 %
  • Lead (Pb):
    max. 5 mg/kg

Specification Test Results

About this item

 :
 :110.98 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903