Order Entry
Austria
ContactUsLinkComponent
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Supplier:  Spectrum Chemical
Certificates
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Viscosity of a 2% Aqueous Solution @ 20D °C:
    3000 - 5600 mPa·s
  • Assay (Methoxy Groups):
    28.0 - 30.0%
  • Assay:Hydroxypropoxy Groups:
    7.0 - 12.0%
  • pH:
    5.0 - 8.0
  • Loss On Drying:
    max. 5.0%

Specification Test Results

Related Information

Frequently Bought Together