Order Entry
Austria
ContactUsLinkComponent
D-(+)-Maltose monohydrate ≥92.0% (by anhydrous basis) NF
D-(+)-Maltose monohydrate ≥92.0% (by anhydrous basis) NF
Supplier:  Spectrum Chemical
Certificates
undefined
D-(+)-Maltose monohydrate ≥92.0% (by anhydrous basis) NF
Supplier:  Spectrum Chemical
Certificates

Specification Test Results

  • Assay (Anhydrous Basis):
    92.0%
  • Water Determination:
    4.5 - 6.5%
  • pH:
    4.0 - 5.5
  • Residue on Ignition:
    Max. 0.05%
  • Dextrin, Starch and Sulfite:
    To Pass Test
  • Identification:
    To Pass Test
  • Residual Solvents:
    To Pass Test
  • Elemental Impurities:
    As Reported

Specification Test Results

Related Information

Frequently Bought Together