Order Entry
Austria
ContactUsLinkComponent
Potassium dihydrogen phosphate ≥98.0% FCC
Potassium dihydrogen phosphate ≥98.0% FCC
Catalog # SPCMP1382-500GM
Supplier:  Spectrum Chemical
CAS Number:  
Potassium dihydrogen phosphate ≥98.0% FCC
Catalog # SPCMP1382-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (Dried Basis):
    98.0 % min.
  • Arsenic (As):
    max.3 mg/kg
  • Fluoride:
    max. 10 mg/kg
  • Insoluble Substances:
    max. 0.2 %
  • Lead (Pb):
    max. 2 mg/kg

Specification Test Results

About this item

 :
 :136.09 g/mol
Storage Temperature:Ambient
CAS Number:7778-77-0
Formula:KH₂PO₄
MDL Number:MFCD00011401