Order Entry
Austria
ContactUsLinkComponent
 
di-Potassium hydrogen phosphate, anhydrous ≥98.0% FCC
Catalog # SPCMPO205-500GM
Supplier:  Spectrum Chemical
CAS Number:  
di-Potassium hydrogen phosphate, anhydrous ≥98.0% FCC
Catalog # SPCMPO205-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (K₂HPO₄):
    98.0 % min.
  • Arsenic (As):
    max.3 mg/kg
  • Fluoride:
    max. 10 mg/kg
  • Insoluble Substances:
    max. 0.2 %
  • Lead (Pb):
    max. 2 mg/kg

Specification Test Results

About this item

 :
 :174.18 g/mol
Storage Temperature:Ambient
CAS Number:7758-11-4
Formula:K₂HPO₄
MDL Number:MFCD00011383