Carbomer 940 resin NF
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:9007-17-4
:9003-01-4
MDL Number:MFCD01941675
Storage Temperature:Ambient
Specification Test Results
- Carboxylic acid groups (dried basis):56.0 - 68.0%
- Viscosity of a 0.5% solution:40,000 - 60,000 mPa
- Loss on drying:2.00%
- Heavy metals:20 µg/g
- Benzene:0.50%
- Identification:Passes test
- Residual solvents:Passes test
Specification Test Results
Related Information
SPCMC1477-100GM_TDS_EN SPCMC1477-2.5KG_TDS_EN SPCMC1477-45KGBL_TDS_EN
Product specification
Frequently Bought Together