Order Entry
Austria
ContactUsLinkComponent
Ammonium thiocyanate ≥97.5% ACS, Sigma-Aldrich®
Irritant
Ammonium thiocyanate ≥97.5% ACS, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Ammonium thiocyanate ≥97.5% ACS, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay:
    Min. 97.5 %
  • pH (5 %, 25 °C):
    4.5 - 6.0
  • Residue on ignition (SO₄):
    Max. 0.025 %
  • Water insoluble matter:
    Max. 0.005 %
  • Iron (Fe):
    Max. 3 ppm
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.005 %
  • Sulfate (SO₄):
    Max. 0.005 %
  • Iodine consuming substances:
    Max. 0.004 meq/g

Specification Test Results

Frequently Bought Together