ContactUsLinkComponent
Manual dispenser and applicator, Scotch-Weld™ EPX™
Manual dispenser and applicator, Scotch-Weld™ EPX™
Catalog # MMMAEPX400H
Supplier:  3M
undefined
Manual dispenser and applicator, Scotch-Weld™ EPX™
Catalog # MMMAEPX400H
Supplier:  3M
Specifications
  • Volume:
    400 ml
  • Colour:
    Grey
  • Description:
    Applicator
  • Pk:
    1
Specifications
About this item
Manual applicator for dispensing EPX adhesive in 38 and 50 ml cartridges supplied with two plungers for 1:1/2:1 and 10:1 adhesive.
  • Dispenses two-component 3M™ Scotch-Weld™ structural adhesive
  • Offers take-it-to-the-job convenience without the need for air hoses or electrical cords
  • Ensures correct mix ratio during dispensing; no need to weigh components
  • Minimises waste and mess associated with hand mixing
  • Reduce plastic waste
Dispense 3M™ Scotch-Weld™ adhesive in duo-pack cartridges with 3M™ Scotch-Weld™ EPX manual applicator. Our applicator mixes and applies two part adhesives. It is commonly used in industrial, metalworking and transportation applications.
Use with both 1:1 and 2:1 200 ml cartridges.