ContactUsLinkComponent
Whatman™ Mini-UniPrep™ Multi-Compressor, Whatman products (Cytiva)
Whatman™ Mini-UniPrep™ Multi-Compressor, Whatman products (Cytiva)
Catalog # WHATMUPMCPBC8
undefined
Whatman™ Mini-UniPrep™ Multi-Compressor, Whatman products (Cytiva)
Catalog # WHATMUPMCPBC8
Specifications
  • Description:
    Mini-UniPrep™ multi compressor with one blue 8 position tray
  • Seq:
    53
  • Pk:
    1
Specifications
About this item
Mini-UniPrep™ multicompressor including one 8-position tray for standard plastic Mini-UniPrep™ filter vials.
  • Streamlines syringeless filtration by filtering up to eight filter vials simultaneously
  • Designed for Cytiva's Mini-UniPrep™ standard plastic filter vial
  • Mini-UniPrep™ vials can be loaded directly into the autosampler
The newly redesigned Whatman™ Mini-UniPrep™ multicompressor streamlines filtration, saves time and reduces hand stress by enabling the filtration of eight Mini-UniPrep™ vials at once.
Ordering information: Additional trays are available, giving the added convenience of pre-loading up to eight Mini-UniPrep™ filter vials.
Delivery: The multiprocessor includes one blue 8-position tray.