ContactUsLinkComponent
Nucleic acid testing for COVID-19
Nucleic acid testing for COVID-19
Catalog # EDVO123
Supplier:  EDVOTEK
undefined
Nucleic acid testing for COVID-19
Catalog # EDVO123
Supplier:  EDVOTEK
Specifications
  • Description:
    COVID
    Corona
    Coronavirus
    Diagnosis
    Electrophoresis
    Pandemic
    COVID-19
    SARS-CoV-2
  • Pk:
    1 KIT
Specifications
About this item
Hands-on simulation of molecular testing for COVID-19.
  • Students learn about characteristics of Coronaviruses
  • Includes all required reagents
  • Use DNA Electrophoresis to analyse simulated samples
  • Activity can be completed in 45 minutes
SARS-CoV-2 is a novel coronavirus that has caused a worldwide outbreak of respiratory disease beginning in 2019.
In this simulated medical test, we will use electrophoresis to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.
Additional items needed but not included: DNA electrophoresis equipment, white-light box, microwave or hot plate.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Store DNA samples in the refrigerator. All other components may be stored at room temperature.