ContactUsLinkComponent
Oxidase, Bactident®, Millipore®
Oxidase, Bactident®, Millipore®
Catalog # 1.00181.0002
Supplier:  Merck
CAS Number:  
Oxidase, Bactident®, Millipore®
Catalog # 1.00181.0002
Supplier:  Merck
CAS Number:  
Specifications
  • Pk:
    50 Tests
  • Enzyme name:
    Oxidase
Specifications
About this item
Oxidase used for the detection of cytochrome oxidase in microorganisms.