ContactUsLinkComponent
Pal spme arrows and kits
Pal spme arrows and kits
Catalog # REST27490
Supplier:  Restek
Pal spme arrows and kits
Catalog # REST27490
Supplier:  Restek
Specifications
  • Description:
    Restek pal spme manual injection kit, Includes spme manual holder; spme manual extraction guide; spme manual injection guide
  • Pk:
    1 KIT
Specifications
About this item
The PAL SPME manual injection kit is designed to house SPME arrows and traditional SPME fibers during extraction and injection steps.
  • Rugged stainless steel construction
  • Longer lifetime
  • Faster extraction
  • Higher sample throughput
  • Better sensitivity allows lower LODs
Due to the relatively large diameter of restek PAL SPME Arrows, you must modify the GC inlet using an instrument-specific conversion kit prior to use.
Accessories information: A GC inlet conversion kit is required in order to use a restek PAL SPME arrow.
Ordering information: These molecular weight ranges are a reasonable approximation; however, end users should verify suitability for their specific application. All restek PAL SPME arrows have 20 mm of phase bonded onto stainless steel.