Order Entry
Austria
ContactUsLinkComponent
Bushnell Haas agar
Bushnell Haas agar
Catalog # HIMEM349-500G
Supplier:  HIMEDIA
CAS Number:  
Bushnell Haas agar
Catalog # HIMEM349-500G
Supplier:  HIMEDIA
CAS Number:  

Specifications

  • Pk:
    500 g
  • Description:
    Bushnell haas agar
  • Seq:
    01
  • Storage temperature:
    <30 °C
  • pH:
    7,0 ±0,2

Specifications

About this item

Bushnell haas agar are recommended for the microbial examination of fuels and for studying microbial hydrocarbon deterioration.

  • White to cream homogeneous free flowing powder
  • Firm gelling, comparable with 2,0% agar gel
  • pH: 6,80 to 7,20
  • Store below 30 °C

Bushnell Haas Agar contains magnesium sulfate (0,200 g/l); calcium chloride (0,020 g/l); monopotassium phosphate (1,000 g/l); dipotassium phosphate (1,000 g/l); ammonium nitrate (1,000 g/l); ferric chloride (0,050 g/l) and agar (20,000 g/l).