ContactUsLinkComponent
Anaerobic basal broth dehydrated culture media
Anaerobic basal broth dehydrated culture media
Catalog # HIMEM1636-500G
Supplier:  HIMEDIA
CAS Number:  
Anaerobic basal broth dehydrated culture media
Catalog # HIMEM1636-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Anaerobic basal broth
Specifications
About this item
Anaerobic basal broth is used for the growth of anaerobic microorganisms, particularly Bacteroides species and other fastidious anaerobes.