Order Entry
Austria
ContactUsLinkComponent
2-Propanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Irritant
Flammable
2-Propanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC323-1
   Safer Human and Environmental Health
CAS Number:  
Irritant
Flammable
2-Propanol ≥99.9%, B&J Brand™ LC-MS for LC-MS, for HPLC, Burdick & Jackson™
Catalog # HONCLC323-1
   Safer Human and Environmental Health
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, GL45
  • Pk:
    6 L
  • Seq:
    5064
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended by default and after discussion
  • Pk Info:
    6x 1 L

Specifications

Specification Test Results

  • GC Analysis:
    99.9 % min.
  • LC-MS Negative Mode (as 4-nitrophenol):
    50 ppb max.
  • LC-MS Positive Mode (as reserpine):
    50 ppb max.
  • Refractive Index (20ºC):
    1.3766 - 1.3778
  • Residue:
    1 mg/L max.
  • Trace Metals:
    Passes test
  • UV Cutoff:
    205 nm max.
  • Water by Karl Fischer Titration:
    0.05 % max.
  • Vapor Pressure (Torr) at20°C:
    32.4

Specification Test Results

About this item

 :
 :60.1 g/mol
Boiling Pt:12.0 %(V)
Melting Pt:–88 °C
Density:0.785 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
 :GHS07
 :GHS02
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674