Bromophenol blue solution for PANPEHA indicator paper, Riedel-de Haën™
Artikelnr. HONC31068-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:669.97 g/mol
Flash Pt:13 °C
Storage Temperature:Ambient
UN:1170
ADR:3,II
CAS Number:115-39-9
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875