ContactUsLinkComponent
Hose with self-closing valve
Hose with self-closing valve
Catalog # HONC49983H-1EA
Hose with self-closing valve
Catalog # HONC49983H-1EA
Specifications
  • Description:
    Hose with self-closing valve
  • Pk:
    1
Specifications
About this item
This hose is suitable for all drums, use in combination with dispensing system or dispensing head.
  • Container accessory
  • For easy and comfortable decanting of solvents