Secondary Drinking Water Metals Certified Reference Material, 500 mg/L Zn, 100 mg/L Cu, 30 mg/L Fe, 5 mg/L Mn in HNO₃ 2%, Fluka™
Catalog # HONCE2165-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Description:Secondary Drinking Water Metals CRM, 500 mg/L Zn, 100 mg/L Cu, 30 mg/L Fe, 5 mg/L Mn in HNO₃ 2%
- UN No.:3264
- Physical form:Liquid
Specifications
About this item
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.