Order Entry
Austria
ContactUsLinkComponent
Primary Drinking Water Metals Certified Reference Material, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%, Fluka™
Primary Drinking Water Metals Certified Reference Material, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%, Fluka™
Catalog # HONCE2164-100ML
CAS Number:  
Primary Drinking Water Metals Certified Reference Material, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%, Fluka™
Catalog # HONCE2164-100ML
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Primary Drinking Water Metals CRM, 100 mg/L Ba, 10 mg/L each of Ag, As, Cr, Hg, Pb, 5 mg/L each of Cd, Se in HNO₃ 2%
  • UN No.:
    3264
  • Physical form:
    Liquid

Specifications

About this item

Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.