About this item
The PURELAB Chorus 2+ range provide >15 MΩ.cm pure water. The systems provide additional bacteria and inorganic quality for sensitive analytical and life science applications above that of basic laboratory work. With its simple design and ease of use, water can be dispensed from the system or from a choice of additional Halo Dispensers.
- Fully Recirculating
- Perfect single system solution for analytical and life science applications requiring >15 MΩ.cm
- Quick and easy replacement of consumables to reduce maintenance times
- Designed to be modular and stackable to save space, whether wall-mounted or under the bench
- Data Capture via USB for system performance validation and software updates
- Multiple dispense
PURELAB Chorus 2+ (RO/DI/UV) provides tap to 15 MΩ.cm pure water for laboratories requiring up to 100 liters per day and is able to fully recirculate to maintain 15 MΩ.cm throughout the day.
The modular nature of PURELAB Chorus 2+ means that your dispense solutions can sit independently from the unit. You can even have the Halo Dispenser installed in an adjacent laboratory. With Halo Dispenser you have the ultimate flexibility.
Suggested applications include electrochemistry, spectrophotometry, feed to ultrapure water, media/buffer preparation, general chemistry.
PURELAB Chorus 2+ (RO/EDI/UV) features our patented recirculating EDI technology: the only EDI system on the market that is able to fully recirculate to maintain >15 MΩ.cm, and is a cost-effective solution for laboratories requiring higher output volumes.
Specifications
- Catalog No:
- ELGAPC220EUBPM1
- ELGAPC210EUBPM1
- ELGAPC220DUBPM1
- Model:
- PURELAB® Chorus 2+ RO/DI/UV
- PURELAB® Chorus 2+ RO/EDI/UV
- PURELAB® Chorus 2+ RO/EDI/UV
- Flow rate:
- >1,5 L/min L/min
- >1,0 L/min L/min
- >1,0 L/min L/min
- Production flow rate:
- 20 L/h
- 10 L/h
- 20 L/h
- TOC content:
- <30 ppb
- <30 ppb
- <30 ppb
- Inorganics @ 25°C:
- 10 to >15 MΩ.cm
- 10 to >15 MΩ.cm
- 10 to >15 MΩ.cm
- Bacteria:
- <0,1 CFU/mL
With POU filter fitted - <0,1 CFU/mL
With POU filter fitted - <0,1 CFU/mL
With POU filter fitted
- Electrical:
- 100 – 240 V
- 100 – 240 V
- 100 – 240 V