Manual crimper for 20 mm crimp vials, level 3, SureSTART™
Supplier: Thermo Fisher Scientific
Certificates
About this item
Use SureSTART™ level 3 manual vial crimpers for 20 mm Crimp vials to ensure that your crimp caps are sealed reliably every time.
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
VWR® Traceable® Digital Thermo-Hygrometer
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
Centrifuge tubes, glass, DURAN®
Supplier: DWK Life Sciences
VWR®, Inserts for Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Manual crimper for 20 mm crimp vials, level 3, SureSTART™
Supplier: Thermo Fisher Scientific
VWR®, Modular Heating Blocks and Accessories for Block Heaters
Supplier: VWR Collection
Stoppers, flat, diamond-ground, DURAN®
Supplier: DWK Life Sciences