HYDRANAL™ - NEXTGEN Coulomat C-FA, Catholyte for Coulometric Karl Fischer Titration, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances)
- Assay of water Max. 30 ppm
- Number of samples Min. 3
Frequently Bought Together