HYDRANAL™ - NEXTGEN solvents for Karl Fischer titration, Fluka™
Catalog # HONC34432-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:1 L
- Description:HYDRANAL™ - NEXTGEN Solvent FI (Methanol based)
- Seq:01
Specifications
About this item
Medium for volumetric two-component Karl Fischer titration (ethanol or methanol based), Free of imidazole (free of CMR substances)