Human Serum AB (Off the Clot)
Catalog # 100-318-100
Supplier: GEMINI BIO-PRODUCTS
Specifications
- Storage temperature:Store frozen at −20…−10 °C
- Pk:100 mL
Specifications
About this item
The off-the-clot human male AB serum is collected from blood that is allowed to coagulate naturally after collection, so there are no anticoagulants or preservatives.
- Recommended for pre-clinical through clinical workflows
- 0,1 µm sterile-filtered
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
Delivery: Transport temperature −70 °C.
Packaging: Shipped on dry ice.