


Acetonitrile, low water content, BioSyn™, meets UV Specifications, Burdick & Jackson™
:75-05-8
:
:H₃CCN
:MFCD00001878
:41.05 g/mol
:82 °C (1.013 hPa)
:–46°C
:0.7857 g/cm³ (20 °C)
:6 °C
:Ambient
:200-835-2
:1648
:3,II
:GHS07
:GHS08
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- GC Analysis:99.9 % min.
- Assay (GC):Min. 99.9 %
- Refractive index (n 20/D):1.3434 - 1.3446
- Water (Karl Fischer):Max. 20 ppm
- Residue from evaporation:Max. 1 mg/l
- Absorbance at 200 nm:Max. 0.05
- Absorbance at 210 nm:Max. 0.04
- Absorbance at 225 nm:Max. 0.01
- UV- cutoff:Max. 190 nm
- VaporPressure(Torr)at20°C:88.8
Frequently Bought Together