EPA vial convenience kits, assembled
Catalog # NSCAB7921
Supplier: THERMO NATIONAL SCIENTIFIC
Specifications
- Cap included:Yes
- Capacity:20,00 ml
- Colour:Brown
- Description:EPA vial convenience kit
- Seq:01
- Environmentally Preferable:
- Pk:100
Specifications
About this item
Thermo Scientific™ national EPA vial convenience kits are designed for discrete water sampling applications.
Kits come with caps, septa and a certificate of analysis.