Order Entry
Austria
Orders LinkContactUsLinkComponent
Sumatriptan Succinate, Reference Standard

Sumatriptan Succinate, Reference Standard

Sumatriptan Succinate, USP reference standard, Vial, 200 mg
Catalog # 1642201.
Supplier:   USP
Sumatriptan Succinate, Reference Standard
Catalog # 1642201.
Supplier:  USP
CAS Number:  
Sumatriptan Succinate, Reference Standard

Sumatriptan Succinate, USP reference standard, Vial, 200 mg

CAS Number:  
Price
539,00 €
Log in for availability
1 * 200 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • Description:
    Sumatriptan Succinate
  • Cat. No.:
    1642201.
  • Formula:
    C₁₄H₂₁N₃O₂S.C₄H₆O₄
  • Description:
    Sumatriptan Succinate
  • CAS No.:
    103628-48-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.