Sumatriptan Related Compounds, Reference Standard
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
3 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1642212.
- 1642223.
- 1642256.
- Description:
- Sumatriptan Succinate Related Compound A
- Sumatriptan Succinate Related Compound C
- Sumatriptan Succinate Related Impurities
- Synonym(s):
- [3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide, succinate salt
- [3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide succinate salt
- CAS No.:
- 545338-89-4
- 103628-48-4
- Formula:
- C₂₇H₃₇N₅O₂S
- C₁₈H₂₇N₃O₆S
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1642212.Sumatriptan Succinate Related Compound ASumatriptan Succinate Related Compound AUSP reference standardVial15 mgLog in for availability1 * 15 mg1 680,00 €Specifications:
More
Specifications
Cat. No.1642212.DescriptionSumatriptan Succinate Related Compound ASynonym(s)[3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide, succinate saltCAS No.545338-89-4FormulaC₂₇H₃₇N₅O₂S - 1642223.Sumatriptan Succinate Related Compound CSumatriptan Succinate Related Compound CUSP reference standardVial50 mgLog in for availability1 * 50 mg1 630,00 €Specifications:
More
Specifications
Cat. No.1642223.DescriptionSumatriptan Succinate Related Compound CSynonym(s)[3-[2-(dimethylamino)ethyl]-1-(hydroxymethyl)-1H-indol-5-yl]-N-methylmethanesulfonamide succinate saltCAS No.Formula - 1642256.Sumatriptan Succinate Related ImpuritiesSumatriptan Succinate Related ImpuritiesUSP reference standardVial25 mgLog in for availability1 * 25 mg1 630,00 €Specifications:
More
Specifications
Cat. No.1642256.DescriptionSumatriptan Succinate Related ImpuritiesSynonym(s)CAS No.103628-48-4FormulaC₁₈H₂₇N₃O₆S



