Order Entry
Austria
ContactUsLinkComponent
Sulfanilamide Melting Point Standard (Approximately 165 degrees), Reference Standard
Sulfanilamide Melting Point Standard (Approximately 165 degrees), Reference Standard
Catalog # 1633007.
Supplier:  USP
CAS Number:  
undefined
Sulfanilamide Melting Point Standard (Approximately 165 degrees), Reference Standard
Catalog # 1633007.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • CAS No.:
    63-74-1
  • Description:
    Sulfanilamide Melting Point Standard
  • Description:
    Sulfanilamide
  • Melting pt.:
    ~165 °C

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard