Order Entry
Austria
ContactUsLinkComponent
Potassium Nitrate Melting Point Standard, Reference Standard
Potassium Nitrate Melting Point Standard, Reference Standard
Catalog # 1548350.
Supplier:  USP
CAS Number:  
Potassium Nitrate Melting Point Standard, Reference Standard
Catalog # 1548350.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    1 g
  • Standards:
    USP reference standard
  • CAS No.:
    7757-79-1
  • Description:
    Potassium Nitrate Melting Point Standard
  • UN No.:
    1486
  • Description:
    Potassium Nitrate
  • Melting pt.:
    ~334 °C

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard